Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Bromo-4-methylpyridine 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 3-Bromo-4-picoline

L19622-06 L19622-03 L19622-14 L19622-06 L19622-03 L19622-14
AAAL19622-14EA 400.99 USD InStock
AAAL19622-14 AAAL19622-03 CAAAAL19622-06 CAAAAL19622-03 CAAAAL19622-14 AAAL19622-06
3-Bromo-4-methylpyridine 98%
3-Bromo-4-methylpyridine


Formula: C₆H₆BrN
MW: 172.02 g/mol
Boiling Pt: 199…200 °C
Density: 1.549
Flash Pt: 79 °C(174 °F)
Storage Temperature: Ambient
MDL Number: MFCD00082592
CAS Number: 3430-22-6
EINECS: 000-000-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty