Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Methoxypyridine 98+%

Supplier: Thermo Scientific Chemicals
Warning

L19625-06 L19625-06 L19625-14 L19625-14
AAAL19625-06EA 79.59 USD InStock
AAAL19625-06 CAAAAL19625-06 AAAL19625-14 CAAAAL19625-14
4-Methoxypyridine 98+%
4-Methoxypyridine


Formula: C₆H₇NO
MW: 109.13 g/mol
Boiling Pt: 191 °C
Melting Pt: 4 °C
Density: 1.075
Flash Pt: 74 °C(165 °F)
Storage Temperature: Ambient
MDL Number: MFCD00674049
CAS Number: 620-08-6
EINECS: 210-624-7
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty