Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-(Methylamino)pyridine 99%

Supplier: Thermo Scientific Chemicals
Warning

L19626-14 L19626-06 L19626-06 L19626-14 L19626-03 L19626-03
AAAL19626-06EA 130.99 USD InStock
AAAL19626-06 CAAAAL19626-06 AAAL19626-14 AAAL19626-03 CAAAAL19626-14 CAAAAL19626-03
4-(Methylamino)pyridine 99%
4-(Methylamino)pyridine


Formula: C₆H₈N₂
MW: 108.14 g/mol
Melting Pt: 124…128 °C
Storage Temperature: Ambient
MDL Number: MFCD00152625
CAS Number: 1121-58-0
ADR: 6.1,III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty