Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Cyanophenylboronic acid 98+%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Dihydroxy(3-cyanophenyl)borane(3-Cyanophenyl)metaboric acid3-Cyanobenzeneboronic acid

L19635-03 L19635-06 L19635-06 L19635-MD L19635-MD L19635-03
AAAL19635-06EA 335.99 USD InStock
CAAAAL19635-MD CAAAAL19635-06 AAAL19635-06 AAAL19635-MD CAAAAL19635-03 AAAL19635-03
3-Cyanophenylboronic acid 98+%
3-Cyanophenylboronic acid


Formula: C₇H₆BNO₂
MW: 146.94 g/mol
Melting Pt: ca 298 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD01318967
CAS Number: 150255-96-2
EINECS: 000-000-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty