Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Quinolinecarbaldehyde 97+%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Quinoline-5-carbaldehydeQuinoline-5-carboxaldehyde

L19641-03 L19641-06 L19641-MD L19641-MD L19641-03 L19641-06
AAAL19641-03EA 182.99 USD InStock
CAAAAL19641-03 CAAAAL19641-06 AAAL19641-03 AAAL19641-06 CAAAAL19641-MD AAAL19641-MD
5-Quinolinecarbaldehyde 97+%
5-Quinolinecarbaldehyde


Formula: C₁₀H₇NO
MW: 157.17 g/mol
Melting Pt: 94…98 °C
Storage Temperature: Ambient
MDL Number: MFCD00805835
CAS Number: 22934-41-4
EINECS: 000-000-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty