Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Iodo-2-furaldehyde 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 5-Iodofurfural

L19670-03 L19670-06 L19670-06 L19670-03
AAAL19670-06EA 166.99 USD InStock
CAAAAL19670-03 AAAL19670-06 CAAAAL19670-06 AAAL19670-03
5-Iodo-2-furaldehyde 97%
5-Iodo-2-furaldehyde


Formula: C₅H₃IO₂
MW: 221.98 g/mol
Melting Pt: 126…127 °C
Storage Temperature: Ambient
MDL Number: MFCD00159503
CAS Number: 2689-65-8
EINECS: 000-000-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty