Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Fluoro-m-anisaldehyde 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 4-Fluoro-3-anisaldehyde4-Fluoro-3-methoxybenzaldehyde

L19671-06 L19671-03 L19671-06 L19671-03
AAAL19671-03EA 42.89 USD InStock
AAAL19671-03 CAAAAL19671-03 AAAL19671-06 CAAAAL19671-06
4-Fluoro-m-anisaldehyde 98%
4-Fluoro-m-anisaldehyde


Formula: C₈H₇FO₂
MW: 154.14 g/mol
Melting Pt: 61…63 °C
Storage Temperature: Ambient
MDL Number: MFCD00143320
CAS Number: 128495-46-5
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty