Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(S)-(-)-1-Boc-3-aminopyrrolidine 95%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: (S)-1-Boc-3-Aminopyrrolidine(S)-(-)-1-BOC-3-aminopyrrolidinetert-butyl (3S)-3-aminopyrrolidine-1-carboxylate

L19696-03 L19696-06 L19696-06 L19696-03
AAAL19696-03EA 109.99 USD InStock
AAAL19696-03 AAAL19696-06 CAAAAL19696-06 CAAAAL19696-03
(S)-(-)-1-Boc-3-aminopyrrolidine 95%
(S)-(-)-1-Boc-3-aminopyrrolidine


Formula: C₉H₁₈N₂O₂
MW: 186.25 g/mol
Boiling Pt: 115 °C/4 mm
Density: 1.020
Storage Temperature: Ambient
MDL Number: MFCD03419271
CAS Number: 147081-44-5
EINECS: 627-302-0
UN: 2810
ADR: "6.1",III
Order Now
Specification Test Results
Optical rotation -3° (neat)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty