Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Bromo-2-nitropyridine 98+%

Supplier: Thermo Scientific Chemicals
Warning

L19748-03 L19748-06 L19748-06 L19748-03
AAAL19748-03EA 34.37 USD InStock
CAAAAL19748-03 CAAAAL19748-06 AAAL19748-03 AAAL19748-06
5-Bromo-2-nitropyridine 98+%
5-Bromo-2-nitropyridine


Formula: C₅H₃BrN₂O₂
Melting Pt: 147…151 °C
Storage Temperature: Ambient
MDL Number: MFCD00160411
CAS Number: 39856-50-3
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty