Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Ethyl-3-methylimidazolium bromide 98+%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 3-Ethyl-1-methyl-1H-imidazol-3-ium bromide

L19761-09 L19761-09 L19761-18 L19761-18
AAAL19761-09EA 64.34 USD InStock
CAAAAL19761-18 AAAL19761-09 CAAAAL19761-09 AAAL19761-18
1-Ethyl-3-methylimidazolium bromide 98+%
1-Ethyl-3-methylimidazolium bromide


Formula: C₆H₁₁BrN₂
MW: 191.07 g/mol
Melting Pt: 70…73 °C
Storage Temperature: Ambient
MDL Number: MFCD03427610
CAS Number: 65039-08-9
EINECS: 000-000-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty