Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Ethyl-3-methylimidazoliumhexafluorophosphate 98+%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 1-Ethyl-3-methyl-1H-imidazol-3-ium hexafluorophosphate(V)

L19762-04 L19762-18 L19762-09 L19762-18 L19762-04 L19762-09
AAAL19762-04EA 62.39 USD InStock
CAAAAL19762-04 CAAAAL19762-18 AAAL19762-04 AAAL19762-09 AAAL19762-18 CAAAAL19762-09
1-Ethyl-3-methylimidazoliumhexafluorophosphate 98+%
1-Ethyl-3-methylimidazolium hexafluorophosphate


Formula: C₆H₁₁F₆N₂P
MW: 256.13 g/mol
Melting Pt: 58…62 °C
Storage Temperature: Ambient
MDL Number: MFCD00216703
CAS Number: 155371-19-0
EINECS: 000-000-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty