Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-(Trifluoromethoxy)phenylboronic acid 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: (3-(Trifluoromethoxy)phenyl)metaboric acid(3-(Trifluoromethoxy)phenyl)boronic acid3-(Trifluoromethoxy)benzeneboronic acidDihydroxy(3-(trifluoromethoxy)phenyl)borane

L19775-03 L19775-06 L19775-03 L19775-06
AAAL19775-03EA 64.59 USD InStock
AAAL19775-03 CAAAAL19775-06 CAAAAL19775-03 AAAL19775-06
3-(Trifluoromethoxy)phenylboronic acid 98%
3-(Trifluoromethoxy)phenylboronic acid


Formula: C₇H₆BF₃O₃
MW: 205.93 g/mol
Melting Pt: ca 80 °C
Storage Temperature: Ambient
MDL Number: MFCD01320697
CAS Number: 179113-90-7
EINECS: 000-000-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty