Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Amino-4-oxocyclohexanecarboxylic acid ethylene ketal 98%

Supplier: Thermo Scientific Chemicals
Synonyms: 8-Amino-1,4-dioxaspiro[4.5]decane-8-carboxylic Acid

L19795-MD L19795-MD L19795-03 L19795-03
AAAL19795-MDEA 63.49 USD InStock
AAAL19795-MD CAAAAL19795-MD AAAL19795-03 CAAAAL19795-03
1-Amino-4-oxocyclohexanecarboxylic acid ethylene ketal 98%
1-Amino-4-oxocyclohexanecarboxylic acid ethylene ketal


Formula: C₉H₁₅NO₄
MW: 201.22 g/mol
Melting Pt: 301…304 °C
Storage Temperature: Ambient
MDL Number: MFCD03790906
CAS Number: 54621-18-0
EINECS: 000-000-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty