Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Dibenzofuranboronic acid 98+%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Dibenzofuran-4-boronic acidDibenzo[b,d]furan-4-ylboronic acid

L19830-03 L19830-06 L19830-03 L19830-06
AAAL19830-06EA 85.19 USD InStock
AAAL19830-06 CAAAAL19830-03 AAAL19830-03 CAAAAL19830-06
4-Dibenzofuranboronic acid 98+%
4-Dibenzofuranboronic acid


Formula: C₁₂H₉BO₃
MW: 212.01 g/mol
Melting Pt: 286…291 °C
Storage Temperature: Ambient
EINECS: 000-000-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty