Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Amino-6-chloropyridine 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 6-Chloropyridin-2-amine6-Chloro-2-pyridylamine

L19836-03 L19836-14 L19836-03 L19836-14 L19836-06 L19836-06
AAAL19836-06EA 92.29 USD InStock
CAAAAL19836-03 CAAAAL19836-14 AAAL19836-06 CAAAAL19836-06 AAAL19836-03 AAAL19836-14
2-Amino-6-chloropyridine 98%
2-Amino-6-chloropyridine


Formula: C₅H₅ClN₂
MW: 128.56 g/mol
Melting Pt: 69…72 °C
Storage Temperature: Ambient
MDL Number: MFCD00234068
CAS Number: 45644-21-1
EINECS: 000-000-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty