Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Aminoisonicotinamide 95%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 2-Aminopyridine-4-carboxamide

L19843-06 L19843-14 L19843-14 L19843-06 L19843-03 L19843-03
AAAL19843-06EA 175.99 USD InStock
CAAAAL19843-06 AAAL19843-06 CAAAAL19843-14 CAAAAL19843-03 AAAL19843-14 AAAL19843-03
2-Aminoisonicotinamide 95%
2-Aminoisonicotinamide


Formula: C₆H₇N₃O
MW: 137.14
Melting Pt: 252…258 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD03791262
CAS Number: 13538-42-6
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty