Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Chloro-2-nitropyridine 97%

Supplier: Thermo Scientific Chemicals
Warning

L19863-06 L19863-MD L19863-MD L19863-06 L19863-03 L19863-03
AAAL19863-06EA 653.99 USD InStock
CAAAAL19863-MD AAAL19863-06 AAAL19863-MD CAAAAL19863-03 AAAL19863-03 CAAAAL19863-06
3-Chloro-2-nitropyridine 97%
3-Chloro-2-nitropyridine


Formula: C₅H₃ClN₂O₂
Melting Pt: 89…92 °C
Storage Temperature: Ambient
MDL Number: MFCD00490110
CAS Number: 54231-32-2
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty