Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-(Cyanomethyl)phenylboronic acid 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 4-(Cyanomethyl)benzeneboronic acid

L19955-06 L19955-03 L19955-06 L19955-03
AAAL19955-06EA 382.99 USD InStock
CAAAAL19955-03 CAAAAL19955-06 AAAL19955-06 AAAL19955-03
4-(Cyanomethyl)phenylboronic acid 98%
4-(Cyanomethyl)phenylboronic acid


Formula: C₈H₈BNO₂
MW: 160.97 g/mol
Melting Pt: ca 196…200 °C
Storage Temperature: Ambient
MDL Number: MFCD01632200
CAS Number: 91983-26-5
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty