Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Nitrophenylboronic acid pinacol ester 98+%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 2-Nitrophenylboronic acid Pinacol ester4,4,5,5-Tetramethyl-2-(2-nitrophenyl)-1,3,2-dioxaborolane2-Nitrobenzeneboronic acid pinacol ester1-Nitro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzene2-Nitrophenylboronic acid, pinacol ester

L19963-06 L19963-03 L19963-06 L19963-03
AAAL19963-06EA 254.99 USD InStock
AAAL19963-06 AAAL19963-03 CAAAAL19963-06 CAAAAL19963-03
2-Nitrophenylboronic acid pinacol ester 98+%
2-Nitrophenylboronic acid pinacol ester


Formula: C₁₂H₁₆BNO₄
MW: 249.07 g/mol
Melting Pt: 42…44 °C
Storage Temperature: Ambient
MDL Number: MFCD02179447
CAS Number: 190788-59-1
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty