Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Octylboronic acid 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: n-Octylboronic acidOctylboronic acid1-n-Octaneboronic acid

L19964-14 L19964-14 L19964-03 L19964-06 L19964-03 L19964-06
AAAL19964-14EA 580.99 USD InStock
CAAAAL19964-06 AAAL19964-14 AAAL19964-03 AAAL19964-06 CAAAAL19964-14 CAAAAL19964-03
1-Octylboronic acid 97%
1-Octylboronic acid


Formula: C₈H₁₉BO₂
MW: 158.05 g/mol
Melting Pt: 81…85 °C
Storage Temperature: Ambient
MDL Number: MFCD01074560
CAS Number: 28741-08-4
EINECS: 000-000-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty