Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-(4-Nitrophenyl)glycerol 99%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: PNPG1-(4-Nitrophenyl)-1,2,3-propanetriol

L19968-MD L19968-06 L19968-03 L19968-06 L19968-03 L19968-MD
AAAL19968-MDEA 65.22 USD InStock
CAAAAL19968-03 AAAL19968-MD AAAL19968-06 CAAAAL19968-MD CAAAAL19968-06 AAAL19968-03
1-(4-Nitrophenyl)glycerol 99%
1-(4-Nitrophenyl)glycerol


Formula: C₉H₁₁NO₅
MW: 213.19 g/mol
Melting Pt: 95…97 °C
Storage Temperature: Ambient
MDL Number: MFCD00017037
CAS Number: 2207-68-3
EINECS: 218-624-9
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty