Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-(2-Fluorophenyl)ethanol 99%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 2-Fluorophenethyl alcohol

L19998-14 L19998-14 L19998-06 L19998-06
AAAL19998-14EA 250.99 USD InStock
CAAAAL19998-14 AAAL19998-14 CAAAAL19998-06 AAAL19998-06
2-(2-Fluorophenyl)ethanol 99%
2-(2-Fluorophenyl)ethanol


Formula: C₈H₉FO
MW: 140.16 g/mol
Boiling Pt: 72 °C/1 mm
Density: 1.045
Flash Pt: 98 °C(208 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002887
CAS Number: 50919-06-7
UN: 2810
ADR: "6.1",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty