Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Chloro-4-(trifluoromethyl)phenylboronic acid 96%

Supplier: Thermo Scientific Chemicals
Warning

L20103-06 L20103-03 L20103-03 L20103-06
AAAL20103-03EA 94.39 USD InStock
CAAAAL20103-03 CAAAAL20103-06 AAAL20103-03 AAAL20103-06
2-Chloro-4-(trifluoromethyl)phenylboronic acid 96%
2-Chloro-4-(trifluoromethyl)phenylboronic acid


Formula: C₇H₅BClF₃O₂
MW: 224.37 g/mol
Melting Pt: 110…112 °C
Storage Temperature: Ambient
MDL Number: MFCD02684337
CAS Number: 254993-59-4
EINECS: 000-000-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty