Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Ethynylaniline 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: (3-Aminophenyl)ethyne(3-Aminophenyl)acetylene1-Amino-3-ethynylbenzene1-Ethynyl-3-aminobenzene

L20124-14 L20124-06 L20124-06 L20124-14
AAAL20124-06EA 84.29 USD InStock
AAAL20124-06 CAAAAL20124-06 AAAL20124-14 CAAAAL20124-14
3-Ethynylaniline 98%
3-Ethynylaniline


Formula: C₈H₇N
MW: 117.15 g/mol
Boiling Pt: 70 °C/1 mm
Melting Pt: 22 °C
Density: 1.040
Flash Pt: >100 °C(212 °F)
Storage Temperature: Ambient
MDL Number: MFCD00014779
CAS Number: 54060-30-9
EINECS: 258-944-6
UN: 1993
ADR: "3",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty