Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3H-1,2-Benzodithiol-one-1,1-dioxide (beaucage reagent) 99%

Supplier: Thermo Scientific Chemicals
Synonyms: 3H-1,2-Benzodithiol-3-one-1,1-dioxide3H-1,2-Benzodithiol-3-one 1,1-Dioxide

L20129-06 L20129-MD L20129-MD L20129-06
AAAL20129-MDEA 41.99 USD InStock
CAAAAL20129-MD CAAAAL20129-06 AAAL20129-MD AAAL20129-06
3H-1,2-Benzodithiol-one-1,1-dioxide (beaucage reagent) 99%
3H-1,2-Benzodithiol-one 1,1-dioxide (Beaucage Reagent)


Formula: C₇H₄O₃S₂
MW: 200.24 g/mol
Melting Pt: 102…107 °C
Storage Temperature: Ambient
UN: 3335
ADR: 9,III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty