Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(1-Propynyl)tributyltin 96%

Supplier: Thermo Scientific Chemicals
Danger

L20201-06 L20201-06 L20201-03 L20201-03
AAAL20201-03EA 60.89 USD InStock
CAAAAL20201-06 AAAL20201-03 CAAAAL20201-03 AAAL20201-06
(1-Propynyl)tributyltin 96%
(1-Propynyl)tributyltin


Formula: C₁₅H₃₀Sn
MW: 329.12
Boiling Pt: 277 °C
Density: 1.082
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD01863650
CAS Number: 64099-82-7
EINECS: 000-000-0
UN: 2788
ADR: "6.1",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty