Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-(Trifluoromethoxy)phenyl isocyanate 97%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 1-Isocyanato-4-(trifluoromethoxy)benzene

B20033-06 B20033-03 B20033-03 B20033-06 B20033-14 B20033-14
AAB20033-03EA 64.21 USD InStock
AAB20033-03 CAAAAB20033-14 CAAAB20033-06 AAB20033-06 CAAAB20033-03 AAAB20033-14
4-(Trifluoromethoxy)phenyl isocyanate 97%
4-(Trifluoromethoxy)phenyl isocyanate


Formula: C₈H₄F₃NO₂
MW: 203.12 g/mol
Boiling Pt: 78 °C/22 mm
Melting Pt: –29 °C
Density: 1.346
Flash Pt: 70 °C(158 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00035702
CAS Number: 35037-73-1
EINECS: 252-328-0
UN: 3390
ADR: "6.1",I
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty