Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Amino-4-methylthiophene-3-carboxamide 98%

Supplier: Thermo Scientific Chemicals

B20043-06 B20043-06 B20043-03 B20043-03
AAB20043-06EA 230.99 USD InStock
CAAAB20043-06 CAAAB20043-03 AAB20043-06 AAB20043-03
2-Amino-4-methylthiophene-3-carboxamide 98%
2-Amino-4-methylthiophene-3-carboxamide


Formula: C₆H₈N₂OS
Melting Pt: 173…175 °C
Storage Temperature: Ambient
MDL Number: MFCD00052594
CAS Number: 4651-97-2
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty