Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Bromo-1-butene 97%

Supplier: Thermo Scientific Chemicals
Danger

B20067-06 B20067-14 B20067-06 B20067-14
AAB20067-14EA 1367.99 USD InStock
CAAAB20067-14 AAB20067-14 CAAAB20067-06 AAB20067-06
2-Bromo-1-butene 97%
2-Bromo-1-butene


Formula: C₄H₇Br
MW: 135 g/mol
Melting Pt: –133.4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00039178
CAS Number: 23074-36-4
EINECS: 245-412-3
UN: 1993
ADR: "3",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty