Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1,3-Propanedisulfonic acid disodium salt 99% (dry weight)

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Sodium propane-1,3-disulphonateSodium propane-1,3-disulfonate

B20092-14 B20092-14 B20092-06 B20092-06
AAB20092-14EA 162.28 USD InStock
AAB20092-14 CAAAB20092-14 CAAAB20092-06 AAB20092-06
1,3-Propanedisulfonic acid disodium salt 99% (dry weight)
1,3-Propanedisulphonic acid disodium salt


Formula: C₃H₆Na₂O₆S₂
MW: 248.19 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00191483
CAS Number: 36589-58-9
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty