Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Amino-3-tert-butyl-1-methylpyrazole 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 3-(tert-Butyl)-1-methyl-1H-pyrazol-5-amine

B20095-03 B20095-14 B20095-14 B20095-06 B20095-06 B20095-03
AAB20095-06EA 271.99 USD InStock
AAB20095-06 AAB20095-03 AAB20095-14 CAAAB20095-06 CAAAB20095-14 CAAAB20095-03
5-Amino-3-tert-butyl-1-methylpyrazole 98%
5-Amino-3-tert-butyl-1-methylpyrazole


Formula: C₈H₁₅N₃
MW: 153.22 g/mol
Melting Pt: 155…158 °C
Storage Temperature: Ambient
MDL Number: MFCD00068002
CAS Number: 118430-73-2
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty