Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Bromo-2-fluorobenzoic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

B20118-14 B20118-06 B20118-06 B20118-14
AAB20118-06EA 95.29 USD InStock
AAB20118-06 AAB20118-14 CAAAB20118-14 CAAAB20118-06
4-Bromo-2-fluorobenzoic acid 98%
4-Bromo-2-fluorobenzoic acid


Formula: C₇H₄BrFO₂
MW: 219.01 g/mol
Melting Pt: 210…213 °C
Storage Temperature: Ambient
MDL Number: MFCD00143263
CAS Number: 112704-79-7
EINECS: 000-000-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty