Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

[2.2]-Paracyclophane 99%

Supplier: Thermo Scientific Chemicals
Synonyms: Tricyclo[8,2,2,24,7]hexadeca-4,6,10,12,13,15-hexaeneTricyclo[8.2.2.2(4,7)]hexadeca-1(12),4,6,10,13,15-hexaene

B20136-03 B20136-06 B20136-06 B20136-03
AAB20136-06EA 203.99 USD InStock
CAAAB20136-03 AAB20136-06 AAB20136-03 CAAAB20136-06
[2.2]-Paracyclophane 99%
[2.2]-Paracyclophane


Formula: C₁₆H₁₆
MW: 208.3 g/mol
Melting Pt: 283…288 °C
Storage Temperature: Ambient
MDL Number: MFCD00003707
CAS Number: 1633-22-3
EINECS: 216-644-2
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty