Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Periodic acid 34% (w/w) in aqueous solution

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Orthoperiodic acidortho-Periodic acid

B20172-0E B20172-36
AAB20172-36EA 139.99 USD InStock
AAB20172-36 AAB20172-0E
Periodic acid 34% (w/w) in aqueous solution
Periodic acid


Formula: H₅IO₆
MW: 227.94 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011344
CAS Number: 10450-60-9
EINECS: 233-937-0
UN: 3098
ADR: "5.1",III
https://us-legacy.vwr.com/stibo/web/std.lang.all/90/77/779077.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty