Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-(3,5-Bis(trifluoromethyl)phenyl)thiourea 98+%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: N-[3,5-Bis(trifluoromethyl)phenyl]thiourea

B20245-03 B20245-06 B20245-03 B20245-06
AAB20245-03EA 44.19 USD InStock
CAAAB20245-03 AAB20245-03 AAB20245-06 CAAAB20245-06
1-(3,5-Bis(trifluoromethyl)phenyl)thiourea 98+%
1-(3,5-Bis(trifluoromethyl)phenyl)thiourea


Formula: C₉H₆F₆N₂S
MW: 288.22
Melting Pt: 168…172 °C
Storage Temperature: Ambient
MDL Number: MFCD00041172
CAS Number: 175277-17-5
UN: 2811
ADR: "6.1",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty