Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Chloro-3-methylhexane 97%

Supplier: Thermo Scientific Chemicals
Warning

B20277-14 B20277-14 B20277-06 B20277-06
AAB20277-06EA 35.89 USD InStock
AAB20277-06 CAAAB20277-06 AAB20277-14 CAAAB20277-14
3-Chloro-3-methylhexane 97%
3-Chloro-3-methylhexane


Formula: C₇H₁₅Cl
MW: 134.65 g/mol
Boiling Pt: 135 °C
Density: 0.879
Flash Pt: 35 °C(95 °F)
Storage Temperature: Ambient
MDL Number: MFCD00060778
CAS Number: 43197-78-0
UN: 1993
ADR: "3",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty