Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Methylthiobenzamide 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Thio-p-toluamide

B20320-03 B20320-03 B20320-06 B20320-06
AAB20320-03EA 57.39 USD InStock
AAB20320-03 CAAAB20320-06 CAAAB20320-03 AAB20320-06
4-Methylthiobenzamide 97%
4-Methylthiobenzamide


Formula: C₈H₉NS
MW: 151.23 g/mol
Melting Pt: 166…169 °C
Storage Temperature: Ambient
MDL Number: MFCD00173750
CAS Number: 2362-62-1
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty