Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Cyclohexylethanol 98%

Supplier: Thermo Scientific Chemicals
Warning

B20363-14 B20363-06 B20363-06 B20363-14
AAB20363-14EA 273.99 USD InStock
AAB20363-14 CAAAB20363-06 AAB20363-06 CAAAB20363-14
1-Cyclohexylethanol 98%
1-Cyclohexylethanol


Formula: C₈H₁₆O
MW: 128.21 g/mol
Boiling Pt: 188…190 °C
Density: 0.92
Flash Pt: 73 °C(163 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001475
CAS Number: 1193-81-3
EINECS: 214-780-7
https://us-legacy.vwr.com/stibo/web/std.lang.all/51/58/2375158.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty