Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium 1-hexadecanesulfonate 97%

Supplier: Thermo Scientific Chemicals
Synonyms: Sodium hexadecane-1-sulphonateHexadecane-1-sulphonic acid sodium saltHexadecane-1-sulfonic acid sodium saltSodium hexadecane-1-sulfonate

B20382-06 B20382-22 B20382-06 B20382-22 B20382-14 B20382-14
AAB20382-06EA 38.69 USD InStock
AAB20382-06 CAAAB20382-06 AAB20382-14 CAAAAB20382-22 CAAAB20382-14 AAAB20382-22
Sodium 1-hexadecanesulfonate 97%
Sodium 1-hexadecanesulphonate


Formula: C₁₆H₃₃NaSO₃
MW: 328.49 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00007528
CAS Number: 15015-81-3
EINECS: 239-100-6
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty