Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Mercaptoacetic acid (thioglycolic acid) 97+%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Mercaptoacetic acid

B20391-22 B20391-36 B20391-0E B20391-22 B20391-0E B20391-36
AAB20391-22EA 49.58 USD InStock
AAB20391-22 CAAAB20391-0E AAB20391-0E AAB20391-36 CAAAB20391-22 CAAAB20391-36
Mercaptoacetic acid (thioglycolic acid) 97+%
Mercaptoacetic acid (Thioglycolic acid)
Reagent that protects tryptophan in amino acid analysis


Formula: HSCH₂COOH
MW: 92.12 g/mol
Boiling Pt: 220 °C
Melting Pt: –12 °C
Density: 1.295
Flash Pt: 119 °C(246 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004876
CAS Number: 68-11-1
EINECS: 200-677-4
UN: 1940
ADR: "8",II
Merck Index: 13,09410
https://us-legacy.vwr.com/stibo/web/std.lang.all/90/10/2299010.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty