Synonyms:
Mercaptoacetic acid
B20391-22
B20391-36
B20391-0E
B20391-22
B20391-0E
B20391-36
AAB20391-22EA
49.58
USD
InStock
AAB20391-22
CAAAB20391-0E
AAB20391-0E
AAB20391-36
CAAAB20391-22
CAAAB20391-36
Mercaptoacetic acid (thioglycolic acid) 97+%
Mercaptoacetic acid (Thioglycolic acid)
Reagent that protects tryptophan in amino acid analysis