Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Chlorobenzonitrile 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: o-Chlorobenzonitrile

B20451-30 B20451-30 B20451-0B B20451-0B
AAB20451-0BEA 172.99 USD InStock
AAB20451-0B CAAAB20451-0B AAB20451-30 CAAAB20451-30
2-Chlorobenzonitrile 98%
2-Chlorobenzonitrile


Formula: C₇H₄ClN
MW: 137.57 g/mol
Boiling Pt: 232 °C
Melting Pt: 43…46 °C
Flash Pt: 122 °C(252 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001779
CAS Number: 873-32-5
EINECS: 212-836-5
ADR: 6.1,III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty