Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Bromo-5-chloropentane 98%

Supplier: Thermo Scientific Chemicals
Warning

B20479-18 B20479-09 B20479-09 B20479-18 B20479-30 B20479-30
AAB20479-18EA 159.99 USD InStock
AAB20479-18 CAAAB20479-30 CAAAB20479-18 CAAAB20479-09 AAB20479-30 AAB20479-09
1-Bromo-5-chloropentane 98%
1-Bromo-5-chloropentane


Formula: BrH₂C(CH₂)₃CH₂Cl
MW: 185.49 g/mol
Boiling Pt: 210…212 °C
Density: 1.408
Flash Pt: 95 °C(203 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001016
CAS Number: 54512-75-3
EINECS: 259-193-7
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty