Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Ethoxy-4-methoxybenzoic acid 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 3-Ethoxy-p-anisic acid

B20489-03 B20489-06 B20489-03 B20489-06
AAB20489-06EA 243.99 USD InStock
CAAAB20489-06 AAB20489-06 AAB20489-03 CAAAB20489-03
3-Ethoxy-4-methoxybenzoic acid 98%
3-Ethoxy-4-methoxybenzoic acid


Formula: C₁₀H₁₂O₃
Melting Pt: 156…160 °C
Storage Temperature: Ambient
MDL Number: MFCD00276591
CAS Number: 2651-55-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty