Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ursodeoxycholic acid 99%

Supplier: Thermo Scientific Chemicals
Synonyms: 3α,7β-Dihydroxy-5β-cholan-24-oic acid 3α,7β-Dihydroxy-5β-cholanic acid 3α,7β-Dihydroxycholan-24-oic acid 3α,7β-Dihydroxycholanic acid UDCA 7β-Hydroxylithocholic acid 7beta-Hydroxylithocholic acid Ursodiol

B20490-14 B20490-06 B20490-03 B20490-03 B20490-06 B20490-14
AAB20490-14EA 459.99 USD InStock
AAB20490-14 AAB20490-03 CAAAB20490-06 CAAAB20490-03 CAAAB20490-14 AAB20490-06
Ursodeoxycholic acid 99%
Ursodeoxycholic acid
A secondary bile acid


Formula: C₂₄H₄₀O₄
MW: 392.58 g/mol
Melting Pt: 201…204 °C
Storage Temperature: Ambient
MDL Number: MFCD00003680
CAS Number: 128-13-2
EINECS: 204-879-3
Merck Index: 12,10026
Order Now

 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty