Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Acetyl-5-bromoindoline 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 1-(5-Bromoindolin-1-yl)ethanone

B20509-06 B20509-06 B20509-14 B20509-03 B20509-14 B20509-03
AAB20509-03EA 49 USD InStock
CAAAB20509-06 AAB20509-03 CAAAAB20509-14 CAAAB20509-03 AAB20509-06 AAAB20509-14
1-Acetyl-5-bromoindoline 98%
1-Acetyl-5-bromoindoline


Formula: C₁₀H₁₀BrNO
MW: 240.1 g/mol
Melting Pt: 118…121 °C
Storage Temperature: Ambient
MDL Number: MFCD00056017
CAS Number: 22190-38-1
EINECS: 000-000-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty