Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2,6-Dichloro-3-nitrotoluene 99%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 1,3-Dichloro-2-methyl-4-nitrobenzene

B20520-18 B20520-09 B20520-06 B20520-09 B20520-18 B20520-06
AAB20520-18EA 148.99 USD InStock
AAB20520-18 CAAAAB20520-06 AAB20520-09 CAAAB20520-09 AAAB20520-06 CAAAB20520-18
2,6-Dichloro-3-nitrotoluene 99%
2,6-Dichloro-3-nitrotoluene


Formula: C₇H₅Cl₂NO₂
MW: 206.03 g/mol
Melting Pt: 53…55 °C
Storage Temperature: Ambient
MDL Number: MFCD00024185
CAS Number: 29682-46-0
EINECS: 249-776-4
UN: 2811
ADR: "6.1",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty