Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Methyl-5-bromo-2-furoate 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Methyl 5-bromofuran-2-carboxylateMethyl 5-Bromo-2-furancarboxylate5-Bromo-2-furoic acid methyll ester5-Bromo-2-furancarboxylic acid methyll esterMethyl 5-Bromo-2-furoate

B20524-06 B20524-03 B20524-03 B20524-06
AAB20524-03EA 65.89 USD InStock
AAB20524-03 AAB20524-06 CAAAB20524-03 CAAAB20524-06
Methyl-5-bromo-2-furoate 97%
Methyl 5-bromo-2-furoate


Formula: C₆H₅BrO₃
MW: 205.01 g/mol
Melting Pt: 62…65 °C
Storage Temperature: Ambient
MDL Number: MFCD00092313
CAS Number: 2527-99-3
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty