Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(±)-1-Hepten-4-ol ≥97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: (±)-Hept-1-en-4-ol

B20591-03 B20591-14 B20591-06 B20591-03 B20591-14 B20591-06
AAB20591-06EA 93.99 USD InStock
CAAAB20591-03 CAAAB20591-14 AAB20591-06 CAAAB20591-06 AAB20591-03 AAB20591-14
(±)-1-Hepten-4-ol ≥97%
(±)-1-Hepten-4-ol


Formula: C₇H₁₄O
MW: 114.19 g/mol
Melting Pt: –61 °C
Storage Temperature: Ambient
MDL Number: MFCD00021933
CAS Number: 3521-91-3
EINECS: 222-535-0
UN: 1987
ADR: "3",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty