Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium deoxycholate monohydrate 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 3α,12α-Dihydroxy-5β-cholan-24-oic acid sodium salt monohydrate 3α,12α-Dihydroxy-5β-cholanic acid sodium salt monohydrate 3α,12α-Dihydroxycholan-24-oic acid sodium salt monohydrate 3α,12α-Dihydroxycholanic acid sodium salt monohydrate Deoxycholic acid sodium salt monohydrate Sodium 3α,12α-dihydroxy-5β-cholanoate monohydrate Sodium 3alpha,12alpha-dihydroxy-5beta-cholanoate monohydrate

B20759-36 B20759-14 B20759-22 B20759-14 B20759-36 B20759-22
AAB20759-22EA 245.23 USD InStock
CAAAB20759-14 AAB20759-22 CAAAB20759-22 AAB20759-14 AAAB20759-36 CAAAAB20759-36
Sodium deoxycholate monohydrate 98%
Sodium deoxycholate monohydrate


Formula: C₂₄H₃₉NaO₄·1H₂O
MW: 432.58 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00150750
CAS Number: 145224-92-6
EINECS: 206-132-7
Order Now

 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty